5-Benzyl-5-azaspiro[2.4]heptan-7-amine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD110237-5GEA 2030 DKK
BLDPBD110237-5G BLDPBD110237-250MG BLDPBD110237-1G
5-Benzyl-5-azaspiro[2.4]heptan-7-amine 98%
5-Benzyl-5-azaspiro[2.4]heptan-7-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₃H₁₈N₂
Molekylvægt: 202,2954 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09607966
CAS-nummer: 129306-07-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR