7,8-Dihydroxy-6-methoxy-2H-chromen-2-one 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD110148-100MGEA 396 DKK
BLDPBD110148-100MG BLDPBD110148-1G BLDPBD110148-250MG
7,8-Dihydroxy-6-methoxy-2H-chromen-2-one 95%
7,8-Dihydroxy-6-methoxy-2H-chromen-2-one

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₈O₅
Molekylvægt: 208,1675 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00006873
CAS-nummer: 574-84-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR