3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD10965-1GEA 194 DKK
BLDPBD10965-1G BLDPBD10965-250MG
3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%
3,5-Bis(trifluoromethyl)-o-phenylenediamine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₈H₆F₆N₂
Molekylvægt: 244,14 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01631430
CAS-nummer: 367-65-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR