2-Amino-3-(6-chloro-1H-indol-3-yl)propanoic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD108918-100MGEA 597 DKK
BLDPBD108918-100MG BLDPBD108918-1G BLDPBD108918-250MG
2-Amino-3-(6-chloro-1H-indol-3-yl)propanoic acid 95%
2-Amino-3-(6-chloro-1H-indol-3-yl)propanoic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₁H₁₁ClN₂O₂
Molekylvægt: 238,6700 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00152036
CAS-nummer: 17808-21-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR