6-Chloro-7-methylchromone 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD108878-250MGEA 504 DKK
BLDPBD108878-250MG BLDPBD108878-5G BLDPBD108878-100MG BLDPBD108878-1G
6-Chloro-7-methylchromone 98%
6-Chloro-7-methylchromone

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₇ClO₂
Molekylvægt: 194,62 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00239401
CAS-nummer: 67029-84-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR