6-Chloro-1H-indole-3-carbonitrile 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD108779-100MGEA 419 DKK
BLDPBD108779-100MG BLDPBD108779-250MG BLDPBD108779-1G
6-Chloro-1H-indole-3-carbonitrile 97%
6-Chloro-1H-indole-3-carbonitrile

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₅ClN₂
Molekylvægt: 176,6024 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07776605
CAS-nummer: 194490-17-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR