Methyl 6-bromo-4-chloroquinoline-2-carboxylate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD108481-250MGEA 349 DKK
BLDPBD108481-250MG BLDPBD108481-5G BLDPBD108481-1G
Methyl 6-bromo-4-chloroquinoline-2-carboxylate 98%
Methyl 6-bromo-4-chloroquinoline-2-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₇BrClNO₂
Molekylvægt: 300,53 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07783793
CAS-nummer: 887587-50-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR