6-Bromo-3-methyl-1H-indazole 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD108447-25GEA 5110 DKK
BLDPBD108447-25G BLDPBD108447-250MG BLDPBD108447-10G BLDPBD108447-5G BLDPBD108447-1G
6-Bromo-3-methyl-1H-indazole 98%
6-Bromo-3-methyl-1H-indazole

Storage condition: Sealed in dry, 2-8°C

Formel: C₈H₇BrN₂
Molekylvægt: 211,0600 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08272244
CAS-nummer: 7746-27-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR