6-Amino-5-nitropyrimidine-2,4(1H,3H)-dione 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD108153-250MGEA 194 DKK
BLDPBD108153-250MG BLDPBD108153-100MG
6-Amino-5-nitropyrimidine-2,4(1H,3H)-dione 97%
6-Amino-5-nitropyrimidine-2,4(1H,3H)-dione

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₄H₄N₄O₄
Molekylvægt: 172,1000 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00603522
CAS-nummer: 3346-22-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR