6,6-Dimethyl-3-(methylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carbonitrile 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD107794-1GEA 1660 DKK
BLDPBD107794-1G BLDPBD107794-250MG BLDPBD107794-100MG
6,6-Dimethyl-3-(methylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carbonitrile 97%
6,6-Dimethyl-3-(methylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carbonitrile

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₁₃NOS₂
Molekylvægt: 251,368 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00084980
CAS-nummer: 175202-50-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR