3-Bromo-5-(methoxycarbonyl)benzoic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD107392-100MGEA 194 DKK
BLDPBD107392-100MG BLDPBD107392-250MG BLDPBD107392-1G
3-Bromo-5-(methoxycarbonyl)benzoic acid 95%
3-Bromo-5-(methoxycarbonyl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₇BrO₄
Molekylvægt: 259,05 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06797598
CAS-nummer: 161796-10-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR