Dimethyl 3-methylpentanedioate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD10728-250MGEA 109 DKK
BLDPBD10728-250MG BLDPBD10728-25G BLDPBD10728-5G BLDPBD10728-1G
Dimethyl 3-methylpentanedioate 95%
Dimethyl 3-methylpentanedioate

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₄O₄
Molekylvægt: 174,19 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00075608
CAS-nummer: 19013-37-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR