Ethyl-4-hydroxy-8-(trifluoromethoxy)quinoline-3-carboxylate 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD1066-5GEA 1240 DKK
BLDPBD1066-5G BLDPBD1066-10G BLDPBD1066-1G BLDPBD1066-25G
Ethyl-4-hydroxy-8-(trifluoromethoxy)quinoline-3-carboxylate 95%
Ethyl-4-hydroxy-8-(trifluoromethoxy)quinoline-3-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₃H₁₀F₃NO₄
Molekylvægt: 301,2180 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00221480
CAS-nummer: 175278-35-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR