3,5-Dimethyl-1H-pyrrole-2-carbaldehyde 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD10598-10GEA 923 DKK
BLDPBD10598-10G BLDPBD10598-250MG BLDPBD10598-5G BLDPBD10598-25G BLDPBD10598-100MG BLDPBD10598-100G BLDPBD10598-1G
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde 98%
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C7H9NO
Molekylvægt: 123,16 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00111522
CAS-nummer: 2199-58-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR