2,6-Dimethyl-3-hydroxypyridine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD10596-250MGEA 85.3 DKK
BLDPBD10596-250MG BLDPBD10596-1G BLDPBD10596-25G BLDPBD10596-10G BLDPBD10596-100G BLDPBD10596-100MG BLDPBD10596-5G
2,6-Dimethyl-3-hydroxypyridine 98%
2,6-Dimethyl-3-hydroxypyridine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₉NO
Molekylvægt: 123,152 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00228692
CAS-nummer: 1122-43-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR