4-Bromo-N-methylbenzenesulphonamide 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD10546-1GEA 77.6 DKK
BLDPBD10546-1G BLDPBD10546-5G
4-Bromo-N-methylbenzenesulphonamide 98%
4-Bromo-N-methylbenzenesulphonamide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₈BrNO₂S
Molekylvægt: 250,1100 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00993162
CAS-nummer: 703-12-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR