(2S,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-carboxylic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD105419-1GEA 4020 DKK
BLDPBD105419-1G BLDPBD105419-250MG BLDPBD105419-100MG
(2S,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-carboxylic acid 97%
(2S,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-carboxylic acid
Formel: C₁₀H₁₁N₅O₅
Molekylvægt: 281,22 g/mol
MDL Number: MFCD00057014
CAS-nummer: 3415-09-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR