4-Chloro-2-methylphenylboronic acid 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD10541-25GEA
512
DKK
BLDPBD10541-25G
BLDPBD10541-100G
BLDPBD10541-10G
BLDPBD10541-5G
BLDPBD10541-1G
4-Chloro-2-methylphenylboronic acid 98%
4-Chloro-2-methylphenylboronic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₇H₈BClO₂ Molekylvægt: 170,40 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02683107 CAS-nummer: 209919-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...