3-Methoxyisonicotinic acid 96%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD10532-5GEA
1070
DKK
BLDPBD10532-5G
BLDPBD10532-250MG
BLDPBD10532-10G
BLDPBD10532-1G
BLDPBD10532-25G
3-Methoxyisonicotinic acid 96%
3-Methoxyisonicotinic acid
Storage condition: Sealed in dry, Room Temperature
Formel:
C₇H₇NO₃ Molekylvægt: 153,1354 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07368621 CAS-nummer: 654663-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...