5-Phenyl-1H-indole 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD105220-100MGEA 985 DKK
BLDPBD105220-100MG BLDPBD105220-1G BLDPBD105220-250MG
5-Phenyl-1H-indole 95%
5-Phenyl-1H-indole

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₄H₁₁N
Molekylvægt: 193,244 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06801971
CAS-nummer: 66616-72-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR