5-Methylisoxazole-4-sulphonyl chloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD104795-1GEA 435 DKK
BLDPBD104795-1G BLDPBD104795-5G BLDPBD104795-250MG
5-Methylisoxazole-4-sulphonyl chloride 97%
5-Methylisoxazole-4-sulphonyl chloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₄H₄ClNO₃S
Molekylvægt: 181,597 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD02681975
CAS-nummer: 321309-26-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR