2-Fluoro-3-nitrotoluene 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD10463-25GEA 187 DKK
BLDPBD10463-25G BLDPBD10463-500G BLDPBD10463-100G BLDPBD10463-10G BLDPBD10463-5G BLDPBD10463-0.5G
2-Fluoro-3-nitrotoluene 98%
2-Fluoro-3-nitrotoluene

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₆FNO₂
Molekylvægt: 155,13 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03412242
CAS-nummer: 437-86-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR