5-Fluoroisophthalaldehyde 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD104087-100MGEA 729 DKK
BLDPBD104087-100MG BLDPBD104087-250MG BLDPBD104087-1G
5-Fluoroisophthalaldehyde 95+%
5-Fluoroisophthalaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₅FO₂
Molekylvægt: 152,12 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08690087
CAS-nummer: 255375-90-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR