5-Chloroisoindolin-1-one 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD103627-100MGEA 256 DKK
BLDPBD103627-100MG BLDPBD103627-5G BLDPBD103627-1G BLDPBD103627-250MG BLDPBD103627-10G
5-Chloroisoindolin-1-one 98%
5-Chloroisoindolin-1-one

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆ClNO
Molekylvægt: 167,592 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08234731
CAS-nummer: 74572-29-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR