5-Chloro-8-nitroquinoline 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD103590-100MGEA 187 DKK
BLDPBD103590-100MG BLDPBD103590-250MG
5-Chloro-8-nitroquinoline 95%
5-Chloro-8-nitroquinoline

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₅ClN₂O₂
Molekylvægt: 208,60 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00552691
CAS-nummer: 6942-98-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR