5-Bromo-N,N-dimethylnicotinamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD103081-1GEA 295 DKK
BLDPBD103081-1G BLDPBD103081-10G BLDPBD103081-5G
5-Bromo-N,N-dimethylnicotinamide 95%
5-Bromo-N,N-dimethylnicotinamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₉BrN₂O
Molekylvægt: 229,0739 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01788370
CAS-nummer: 292170-96-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR