5-Amino-2-chlorobenzonitrile 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD102142-1GEA
77.6
DKK
BLDPBD102142-1G
BLDPBD102142-5G
BLDPBD102142-10G
BLDPBD102142-25G
BLDPBD102142-100G
BLDPBD102142-250MG
5-Amino-2-chlorobenzonitrile 97%
5-Amino-2-chlorobenzonitrile
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formel:
C₇H₅ClN₂ Molekylvægt: 152,5800 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00800459 CAS-nummer: 35747-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...