5-Amino-1-(4-methoxyphenyl)-1H-pyrazole-4-carboxylic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD102025-1GEA 551 DKK
BLDPBD102025-1G BLDPBD102025-5G BLDPBD102025-250MG
5-Amino-1-(4-methoxyphenyl)-1H-pyrazole-4-carboxylic acid 95%
5-Amino-1-(4-methoxyphenyl)-1H-pyrazole-4-carboxylic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₁H₁₁N₃O₃
Molekylvægt: 233,22 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00973939
CAS-nummer: 14678-95-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR