5-Acetylpicolinonitrile 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD101928-250MGEA 962 DKK
BLDPBD101928-250MG BLDPBD101928-1G BLDPBD101928-100MG
5-Acetylpicolinonitrile 98%
5-Acetylpicolinonitrile

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₆N₂O
Molekylvægt: 146,14 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06637533
CAS-nummer: 249583-84-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR