5-(4-Isopropylphenyl)-1,3,4-thiadiazol-2-amine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD101040-1GEA 109 DKK
BLDPBD101040-1G BLDPBD101040-5G
5-(4-Isopropylphenyl)-1,3,4-thiadiazol-2-amine 95%
5-(4-Isopropylphenyl)-1,3,4-thiadiazol-2-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₁H₁₃N₃S
Molekylvægt: 219,3100 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD02032690
CAS-nummer: 100987-89-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR