5-(4-Fluorophenyl)oxazole 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD101022-250MGEA 357 DKK
BLDPBD101022-250MG BLDPBD101022-100MG BLDPBD101022-10G BLDPBD101022-1G BLDPBD101022-5G
5-(4-Fluorophenyl)oxazole 97%
5-(4-Fluorophenyl)oxazole

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₆FNO
Molekylvægt: 163,1485 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00085087
CAS-nummer: 128101-19-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR