(S)-2-Cbz-aminobutane-1,4-diol 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD0899-1GEA 590 DKK
BLDPBD0899-1G BLDPBD0899-5G BLDPBD0899-25G
(S)-2-Cbz-aminobutane-1,4-diol 97%
(S)-2-Cbz-aminobutane-1,4-diol

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₂H₁₇NO₄
Molekylvægt: 239,2700 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03426373
CAS-nummer: 118219-23-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR