3-Formyl-6-methylchromone 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD0707-25GEA 2050 DKK
BLDPBD0707-25G BLDPBD0707-1G BLDPBD0707-250MG BLDPBD0707-5G
3-Formyl-6-methylchromone 95%
3-Formyl-6-methylchromone

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₈O₃
Molekylvægt: 188,1794 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00138943
CAS-nummer: 42059-81-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR