Ethyl 3-(3-bromophenyl)-3-oxopropanoate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD0501-5GEA 1140 DKK
BLDPBD0501-5G BLDPBD0501-10G BLDPBD0501-1G BLDPBD0501-250MG BLDPBD0501-25G
Ethyl 3-(3-bromophenyl)-3-oxopropanoate 95%
Ethyl 3-(3-bromophenyl)-3-oxopropanoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₁BrO₃
Molekylvægt: 271,1100 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03424812
CAS-nummer: 21575-91-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR