3-Amino-1-Boc-piperidine 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD0336-1GEA
46.6
DKK
BLDPBD0336-1G
BLDPBD0336-25G
BLDPBD0336-5G
BLDPBD0336-500G
BLDPBD0336-10G
BLDPBD0336-100G
3-Amino-1-Boc-piperidine 97%
3-Amino-1-Boc-piperidine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₁₀H₂₀N₂O₂ Molekylvægt: 200,28 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01861219 CAS-nummer: 184637-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...