2,2'-((2-Nitro-1,4-phenylene)bis(azanediyl))diethanol 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD0275-10GEA 225 DKK
BLDPBD0275-10G BLDPBD0275-1G BLDPBD0275-250MG BLDPBD0275-25G BLDPBD0275-5G
2,2'-((2-Nitro-1,4-phenylene)bis(azanediyl))diethanol 97%
2,2'-((2-Nitro-1,4-phenylene)bis(azanediyl))diethanol

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₀H₁₅N₃O₄
Molekylvægt: 241,2400 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00239474
CAS-nummer: 84041-77-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR