4,4,5,5-Tetramethyl-2-(3-(neopentyloxy)phenyl)-1,3,2-dioxaborolane 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01591060-1GEA 667 DKK
BLDPBD01591060-1G BLDPBD01591060-250 BLDPBD01591060-5G
4,4,5,5-Tetramethyl-2-(3-(neopentyloxy)phenyl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(3-(neopentyloxy)phenyl)-1,3,2-dioxaborolane

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₇H₂₇BO₃
Molekylvægt: 290,2055 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2246560-92-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR