Ethyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01573572-250EA 311 DKK
BLDPBD01573572-250 BLDPBD01573572-5G BLDPBD01573572-1G
Ethyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 95%
Ethyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₆H₂₃BO₄
Molekylvægt: 290,16 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 1150271-63-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR