N-Benzyl-2-bromo-4-methylbenzenesulphonamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01569329-1GEA 1720 DKK
BLDPBD01569329-1G BLDPBD01569329-250 BLDPBD01569329-5G
N-Benzyl-2-bromo-4-methylbenzenesulphonamide 95%
N-Benzyl-2-bromo-4-methylbenzenesulphonamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₄BrNO₂S
Molekylvægt: 340,2355 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12651434
CAS-nummer: 1184443-16-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR