7-Chloro-3-iodoquinoline 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01547973-100EA 303 DKK
BLDPBD01547973-100 BLDPBD01547973-1G BLDPBD01547973-250
7-Chloro-3-iodoquinoline 95%
7-Chloro-3-iodoquinoline

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₉H₅ClIN
Molekylvægt: 289,5002 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD28667978
CAS-nummer: 1424245-91-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR