2-Bromo-3,4-dichloro-1-methoxybenzene 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01547193-100EA 590 DKK
BLDPBD01547193-100 BLDPBD01547193-1G BLDPBD01547193-250
2-Bromo-3,4-dichloro-1-methoxybenzene 95%
2-Bromo-3,4-dichloro-1-methoxybenzene

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₅BrCl₂O
Molekylvægt: 255,9240 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD31725577
CAS-nummer: 174913-13-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR