N1,N6-Dibutyladipamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01543411-5GEA 1240 DKK
BLDPBD01543411-5G BLDPBD01543411-1G
N1,N6-Dibutyladipamide 95%
N1,N6-Dibutyladipamide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₄H₂₈N₂O₂
Molekylvægt: 256,38 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03405231
CAS-nummer: 10472-27-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR