2-((4-(Trifluoromethoxy)phenyl)amino)thiazole-5-carboxylic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01543375-100EA 1200 DKK
BLDPBD01543375-100 BLDPBD01543375-1G BLDPBD01543375-250
2-((4-(Trifluoromethoxy)phenyl)amino)thiazole-5-carboxylic acid 95%
2-((4-(Trifluoromethoxy)phenyl)amino)thiazole-5-carboxylic acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₁H₇F₃N₂O₃S
Molekylvægt: 304,24 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 1500622-59-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR