2-(5-Ethoxy-2-fluoro-3-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01542318-5GEA 2340 DKK
BLDPBD01542318-5G BLDPBD01542318-250 BLDPBD01542318-1G
2-(5-Ethoxy-2-fluoro-3-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(5-Ethoxy-2-fluoro-3-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₅H₁₉BF₄O₃
Molekylvægt: 334,11 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 2828446-36-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR