Ethyl 4-(3-chloro-6-fluorobenzo[b]thiophene-2-carbonyl)piperazine-1-carboxylate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01527733-50MEA 12070 DKK
BLDPBD01527733-50M BLDPBD01527733-10M BLDPBD01527733-25M
Ethyl 4-(3-chloro-6-fluorobenzo[b]thiophene-2-carbonyl)piperazine-1-carboxylate 95%
Ethyl 4-(3-chloro-6-fluorobenzo[b]thiophene-2-carbonyl)piperazine-1-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₆H₁₆ClFN₂O₃S
Molekylvægt: 370,82 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04064660
CAS-nummer: 620570-09-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR