1-(Furan-2-yl)-N,N-dimethylmethanamine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01520470-1GEA 4880 DKK
BLDPBD01520470-1G BLDPBD01520470-100 BLDPBD01520470-250
1-(Furan-2-yl)-N,N-dimethylmethanamine 95%
1-(Furan-2-yl)-N,N-dimethylmethanamine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₁₁NO
Molekylvægt: 125,16 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01365940
CAS-nummer: 14496-34-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR