3-Chloro-6-iodopyridazine 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD01520063-25GEA
783
DKK
BLDPBD01520063-25G
BLDPBD01520063-1G
BLDPBD01520063-100
BLDPBD01520063-5G
3-Chloro-6-iodopyridazine 95%
3-Chloro-6-iodopyridazine
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formel:
C₄H₂ClIN₂ Molekylvægt: 240,43 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD08275187 CAS-nummer: 135034-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...