N-(5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)propan-2-amine hydrochloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01513748-5GEA 4110 DKK
BLDPBD01513748-5G BLDPBD01513748-250 BLDPBD01513748-1G
N-(5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)propan-2-amine hydrochloride 95%
N-(5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)propan-2-amine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₆H₂₆BCl₂NO₂
Molekylvægt: 346,1001 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 2828445-82-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR