2-Oxa-7-azaspiro[4.4]nonane hemioxalate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01512687-50MEA 745 DKK
BLDPBD01512687-50M BLDPBD01512687-100 BLDPBD01512687-250
2-Oxa-7-azaspiro[4.4]nonane hemioxalate 95%
2-Oxa-7-azaspiro[4.4]nonane hemioxalate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₆H₂₈N₂O₆
Molekylvægt: 344,4033 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2828444-89-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR