(2S,4R)-4-Fluoro-2-methylpyrrolidine hydrochloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD01510327-250EA 5790 DKK
BLDPBD01510327-250 BLDPBD01510327-100 BLDPBD01510327-50M
(2S,4R)-4-Fluoro-2-methylpyrrolidine hydrochloride 95%
(2S,4R)-4-Fluoro-2-methylpyrrolidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₁₁ClFN
Molekylvægt: 139,5989 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 2828444-35-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR